Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBCK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA573012
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA573012 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA573012 Supplier Invitrogen™ Supplier No. PA573012
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Orangutan/Chimpanzee (100%), New world monkey (89%), Bovine/Guinea Pig/Mole Rat (83%), Canine/Elephant/Rabbit (78%), Panda/Oppossum (67%), Rat (61%).

RBCK1 encodes a protein that is similar to mouse UIP28/UbcM4 interacting protein. Alternative splicing has been observed at this locus, resulting in distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBCK1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene RBCK1
Gene Accession No. Q9BYM8, Q9WUB0
Gene Alias AL033326; C20orf18; HBV-associated factor 4; heme-oxidized IRP2 ubiquitin ligase 1; Heme-oxidized IRP2 ubiquitin ligase 1 homolog; hepatitis B virus X-associated protein 4; HOIL1; HOIL-1; HOIL-1L; novel zinc finger, C3HC4 type (RING finger) protein; PBMEI; PGBM1; Pkcbpb15; protein kinase C-binding protein Beta15; protein kinase C-binding protein beta-15; RANBP2-type and C3HC4-type zinc finger containing 1; RanBP-type and C3HC4-type zinc finger containing 1; ranBP-type and C3HC4-type zinc finger-containing protein 1; RBCC protein interacting with PKC; RBCC protein interacting with PKC 1; RBCC protein interacting with PKC1; Rbck; RBCK1; RBCK2; RING finger protein 54; RING-type E3 ubiquitin transferase HOIL-1; RNF54; UBCE7IP3; ubcM4-interacting protein 28; ubiquitin conjugating enzyme 7 interacting protein 3; ubiquitin-conjugating enzyme 7-interacting protein 3; Uip28; XAP3; XAP4; ZRANB4
Gene Symbols RBCK1
Host Species Rabbit
Immunogen A portion of amino acids 150-200 from human RBCk1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10616, 24105
Target Species Human, Mouse, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.