Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBCK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115828
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115828 Supplier Invitrogen™ Supplier No. PA5115828
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RBCK1.

RBCK1 encodes a protein that is similar to mouse UIP28/UbcM4 interacting protein. Alternative splicing has been observed at this locus, resulting in distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBCK1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene RBCK1
Gene Accession No. Q62921, Q9BYM8, Q9WUB0
Gene Alias AL033326; C20orf18; HBV-associated factor 4; heme-oxidized IRP2 ubiquitin ligase 1; Heme-oxidized IRP2 ubiquitin ligase 1 homolog; hepatitis B virus X-associated protein 4; HOIL1; HOIL-1; HOIL-1L; novel zinc finger, C3HC4 type (RING finger) protein; PBMEI; PGBM1; Pkcbpb15; protein kinase C-binding protein Beta15; protein kinase C-binding protein beta-15; RANBP2-type and C3HC4-type zinc finger containing 1; RanBP-type and C3HC4-type zinc finger containing 1; ranBP-type and C3HC4-type zinc finger-containing protein 1; RBCC protein interacting with PKC; RBCC protein interacting with PKC 1; RBCC protein interacting with PKC1; Rbck; RBCK1; RBCK2; RING finger protein 54; RING-type E3 ubiquitin transferase HOIL-1; RNF54; UBCE7IP3; ubcM4-interacting protein 28; ubiquitin conjugating enzyme 7 interacting protein 3; ubiquitin-conjugating enzyme 7-interacting protein 3; Uip28; XAP3; XAP4; ZRANB4
Gene Symbols RBCK1
Host Species Rabbit
Immunogen A synthesized peptide derived from human RBCK1(Accession Q9BYM8), corresponding to amino acid residues K477-H510.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10616, 24105, 60383
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.