Learn More
RBEL1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303726A

Description
The recommended shelf life for this product is 1 year from date of receipt.
May enhance cellular proliferation. May reduce growth inhibitory activity of CDKN2A.Specifications
RBEL1 | |
Polyclonal | |
Unconjugated | |
RABL6 | |
PARF, RBEL1, pp8875, Rab-like GTP-binding protein 1C, partner of ARF, putative GTP-binding protein Parf | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55684 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q3YEC7 | |
RABL6 | |
Between 525 and 575 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.