Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBFA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559196
Description
Immunogen sequence: SKVSLTPDFS ACRAYWKTTL SAEQNAHMEA VLQRSAAHMR HLLMSQQTLR NVPPIVFVQD KGNAALAELD QLLAVADFGP RDERDNFVQN DFR Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 74%.
RBFA is a protein coding gene and also known as Ribosome Binding Factor A.
Specifications
RBFA | |
Polyclonal | |
Unconjugated | |
RBFA | |
1110032A13Rik; AI595940; C18orf22; HsT169; Putative ribosome-binding factor A, mitochondrial; Rbfa; RGD1311910; ribosome binding factor A; ribosome binding factor A (putative) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79863 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N0V3 | |
RBFA | |
Recombinant protein corresponding to Human RBFA. Recombinant protein control fragment (Product #RP-98237). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction