Learn More
Invitrogen™ RBFOX1 Monoclonal Antibody (OTI1F7)

Description
RBFOX1 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | RBFOX1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1F7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Rbfox1 |
| Gene Accession No. | Q9NWB1 |
| Gene Alias | 2BP1; A FOX-1; A2BP; A2BP1; ataxin 2 binding protein 1; ataxin 2-binding protein 1; ataxin-2-binding protein 1; FOX1; fox-1; fox-1 homolog A; fox-1-like RNA-binding protein 1; hexaribonucleotide binding protein 1; hexaribonucleotide binding protein 1 isoform alpha; hexaribonucleotide binding protein 1 isoform beta; hexaribonucleotide-binding protein 1; HRNBP1; MNCb-3035; RBFOX1; RNA binding fox-1 homolog 1; RNA binding protein fox-1 homolog 1; RNA binding protein, fox-1 homolog (C. elegans) 1; RNA binding protein, fox-1 homolog 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.