Learn More
Invitrogen™ RBFOX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114130
Description
Immunogen sequence: TAAAYSDRNQ FVFVAADEIS CNTSAVTDEF MLPTPTTTHL LQPPPTALVP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Zebrafish: 100%.
Ataxin-2 binding protein 1 has an RNP motif that is highly conserved among RNA-binding proteins. This protein binds to the C-terminus of ataxin-2 and may contribute to the restricted pathology of spinocerebellar ataxia type 2 (SCA2). Ataxin-2 is the gene product of the SCA2 gene which causes familial neurodegenerative diseases. Ataxin-2 binding protein 1 and ataxin-2 are both localized in the trans-Golgi network. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene. Additional transcript variants have been found but their full length nature has not been determined.
Specifications
| RBFOX1 | |
| Polyclonal | |
| Unconjugated | |
| Rbfox1 | |
| 2BP1; A FOX-1; A2BP; A2BP1; ataxin 2 binding protein 1; ataxin 2-binding protein 1; ataxin-2-binding protein 1; FOX1; fox-1; fox-1 homolog A; fox-1-like RNA-binding protein 1; hexaribonucleotide binding protein 1; hexaribonucleotide binding protein 1 isoform alpha; hexaribonucleotide binding protein 1 isoform beta; hexaribonucleotide-binding protein 1; HRNBP1; MNCb-3035; RBFOX1; RNA binding fox-1 homolog 1; RNA binding protein fox-1 homolog 1; RNA binding protein, fox-1 homolog (C. elegans) 1; RNA binding protein, fox-1 homolog 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 54715 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NWB1 | |
| Rbfox1 | |
| Synthetic peptide directed towards the middle region of human A2BP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.