Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBFOX2 Monoclonal Antibody (1E7G10)
GREENER_CHOICE

Catalog No. PIMA538623
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538623 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538623 Supplier Invitrogen™ Supplier No. MA538623
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody has been tested in indirect ELISA.

This gene is one of several human genes similar to the C. elegans gene Fox-1. This gene encodes an RNA binding protein that is thought to be a key regulator of alternative exon splicing in the nervous system and other cell types. The protein binds to a conserved UGCAUG element found downstream of many alternatively spliced exons and promotes inclusion of the alternative exon in mature transcripts. The protein also interacts with the estrogen receptor 1 transcription factor and regulates estrogen receptor 1 transcriptional activity. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBFOX2
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1E7G10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene RBFOX2
Gene Accession No. O43251
Gene Alias dJ106I20.3; Fbm2; fox-1 homolog B; Fox-1 homolog Fxh; fox-1 homologue; FOX2; Fox-2; Fxh; Fyn-binding molecule 2; hexaribonucleotide-binding protein 2; HNRBP2; Hrnbp2; Rbfox2; Rbm9; repressor of tamoxifen transcriptional activity; RNA binding fox-1 homolog 2; RNA binding motif protein 9; RNA binding protein fox-1 homolog 2; RNA binding protein fox-1 homolog 2; RNA binding protein, fox-1 homolog (C. elegans) 2; RNA binding protein, fox-1 homolog (C. elegans) 2; RNA binding protein, fox-1 homolog 2; RNA-binding motif protein 9; RNA-binding protein 9; RTA; zgc:85694
Gene Symbols RBFOX2
Host Species Mouse
Immunogen Purified recombinant fragment of human RBFOX2 (aa: 1-145) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23543
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.