Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NLRP3/NALP3 Antibody (25N10E9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$192.47 - $443.24
Specifications
| Antigen | NLRP3/NALP3 |
|---|---|
| Clone | 25N10E9 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2-5 ug/ml, Simple Western 1:50 |
| Applications | Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP203948SS
![]() |
Novus Biologicals
NBP203948SS |
0.025 mg |
Each for $192.47
|
|
|||||
NBP203948
![]() |
Novus Biologicals
NBP203948 |
0.1 mg |
Each for $443.24
|
|
|||||
Description
NLRP3/NALP3 Monoclonal specifically detects NLRP3/NALP3 in Human, Mouse samples. It is validated for Western Blot, Simple Western.Specifications
| NLRP3/NALP3 | |
| 1.0 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Alzheimers Research, Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Inflammation, Neurodegeneration, Signal Transduction | |
| NP_001073289 | |
| 114548 | |
| A recombinant protein corresponding to the NOD domain of murine NLRP3/NALP3 Antibody (25N10E9) (amino acids 216-385) was used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 25N10E9 | |
| Western Blot 2-5 ug/ml, Simple Western 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, AVP, C1orf7, Caterpiller protein 1.1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, NLRP3 monoclonal, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1 | |
| NLRP3 | |
| IgG2b κ | |
| Protein G purified | |
| The epitope that 25N10E9 binds to the NOD region of NLRP3/NALP3 has not been mapped. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title