Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBFOX3/NeuN Antibody, Novus Biologicals™

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NBP2104910.5ML
Description
RBFOX3/NeuN Polyclonal specifically detects RBFOX3/NeuN in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RBFOX3/NeuN | |
Polyclonal | |
Unconjugated | |
10 mM PBS (0.9% isotonic, w/v, pH 7.2). with 0.02% sodium azide | |
RBFOX3 | |
Hens were immunized with a synthetic peptide-keyhole limpet hemocyanin (KLH) conjugate. This synthetic peptide corresponded to the Fox-3 gene product (also known as the Neu-N antigen), but was shared between the human NP_001076044, NCBI) and mouse (NP_001020102, NCBI) sequences. After repeated injections into the hens, immune eggs were collected, and the IgY fractions were purified from the yolks. | |
Primary | |
Human, Mouse | |
IgY |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.10 mg/mL | |
Immunohistochemistry 1:1000-1:2000, Immunocytochemistry/Immunofluorescence 1:1000-1:2000, Immunohistochemistry-Paraffin 1:1000-1:2000 | |
FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, neuronal nuclei, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3 | |
Chicken | |
0.5 mL | |
146713 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction