Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBG1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570753
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570753 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570753 Supplier Invitrogen™ Supplier No. PA570753
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 86%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 100%; Rat: 100%.

RABGAP1L is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBG1L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Rabgap1l
Gene Accession No. B7ZAP0, Q5R372
Gene Alias 5830411O09Rik; 8430421H08Rik; 9630005B12Rik; AW049894; expressed in hematopoietic cells, heart, liver (HLL); Hh1; Hhl; Kiaa0471; mKIAA0471; RAB GTPase activating protein 1 like; RAB GTPase activating protein 1-like; rab GTPase-activating protein 1-like; Rab GTPase-activating protein 1-like, isoform 10; RAB GTPase-activating protein 1-like-like protein; rab6 GTPase activating protein-like; RABGAP1L; RGD1304620; TBC1 domain family, member 18; TBC1D18
Gene Symbols Rabgap1l
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human RBG1L.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9910
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.