Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBJ (DNAJC27) Monoclonal Antibody (OTI10A9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF811078


Description
RBJ (DNAJC27) Monoclonal antibody specifically detects RBJ (DNAJC27) in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
RBJ (DNAJC27) | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q08849, Q13277, Q64704 | |
Mouse | |
Protein A/G | |
RUO | |
20908, 6809, 81802 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI10A9 | |
Unconjugated | |
Stx3 | |
AI639580; APAH1; C330021A05Rik; DnaJ (Hsp40) homolog, subfamily C, member 27; DnaJ heat shock protein family (Hsp40) member C27; dnaJ homolog subfamily C member 27; dnajc27; LOW QUALITY PROTEIN: dnaJ homolog subfamily C member 27; rab and DnaJ domain containing; rab and DnaJ domain-containing protein; Rabj; RabJS; Rab-related GTP-binding protein; Ras-associated protein Rap1; RBJ; zgc:194263; zgc:194279 | |
Full length human recombinant protein of human STX3 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction