Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBKS Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297078G
Description
RBKS Polyclonal specifically detects RBKS in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RBKS | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
EC 2.7.1.15, RBSKDKFZp686G13268,5230400M11Rik, ribokinase | |
Produced in rabbits immunized with purified, recombinant RBKS (NP_071411.1; Met1-Phe322). | |
0.1 mL | |
metabolism | |
64080 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction