Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBKS Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297079MFV610
Description
RBKS Polyclonal antibody specifically detects RBKS in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
RBKS | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry-Paraffin | |
EC 2.7.1.15, RBSKDKFZp686G13268,5230400M11Rik, ribokinase | |
Rabbits immunized with purified, recombinant Human RBKS (Accession#: NP_071411.1, Met1-Phe322) | |
0.1 mL | |
metabolism | |
64080 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction