Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBKS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582693
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582693 Supplier Invitrogen™ Supplier No. PA582693
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CQVVIITLGA EGCVVLSQTE PEPKHIPTEK VKAVDTTGAG DSFVGALAFY LAYYPNLSLE DMLNRSNFIA AVSVQAAGTQ SSYPYKKDLP LTLF Highest antigen sequence indentity to the following orthologs: Mouse - 89%, Rat - 87%.

RBSK belongs to the pfkB family of carbohydrate kinases. It phosphorylates ribose to form ribose-5-phosphate in the presence of ATP and magnesium as a first step in ribose metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBKS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RBKS
Gene Accession No. Q9H477
Gene Alias 5230400M11Rik; RBKS; RBSK; Ribokinase; RK
Gene Symbols RBKS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RBKS. Recombinant protein control fragment (Product #RP-91761).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64080
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.