Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBL1 Monoclonal Antibody (OTI4E1)
GREENER_CHOICE

Catalog No. PIMA527449
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527449 Supplier Invitrogen™ Supplier No. MA527449
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RBL1 Monoclonal Antibody for Western Blot

RB-like 1 is similar in sequence and possibly function to the product of the retinoblastoma 1 (RB1) gene. The RB1 gene product is a tumor suppressor protein that appears to be involved in cell cycle regulation, as it is phosphorylated in the S to M phase transition and is dephosphorylated in the G1 phase of the cell cycle. Both the RB1 protein and RB-like 1 can form a complex with adenovirus E1A protein and SV40 large T-antigen, with the SV40 large T-antigen binding only to the unphosphorylated form of each protein. In addition, both proteins can inhibit the transcription of cell cycle genes containing E2F binding sites in their promoters. Due to the sequence and biochemical similarities with the RB1 protein, it is thought that RB-like 1 may also be a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBL1
Applications Western Blot
Classification Monoclonal
Clone OTI4E1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RBL1
Gene Accession No. P28749
Gene Alias 107 kDa retinoblastoma-associated protein; AW547426; CP107; DNA-binding protein RFX5; MGC40006; p107; pRb1; RB transcriptional corepressor like 1; Rbl1; Regulatory factor X 5; regulatory factor X, 5 (influences HLA class II expression); regulatory factor X5; retinoblastoma-like 1; retinoblastoma-like 1 (p107); retinoblastoma-like protein 1; RFX5
Gene Symbols RBL1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 739-1014 of RBL1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5933
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.