Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566080
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566080 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566080 Supplier Invitrogen™ Supplier No. PA566080
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EKCFKCGVPKS EAEQKLPLGT RLDQQTLPLG GRELSQGLLP LPQPYQAQGV LASQALSQGS EPSSENANDT IILRNLNPHS T Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

The protein encoded by this gene contains RNA recognition motif found in a variety of RNA binding proteins, including various hnRNP proteins, proteins implicated in regulation of alternative splicing, and protein components of snRNPs. In vitro studies showed that the rat homolog bound to RNA homopolymers, with a preference for G and U polyribonucleotides. This gene is part of a gene cluster on chromosome Xp11.23, and its 3' end lies within 20 kb upstream of UBE1. Two transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM10
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RBM10
Gene Accession No. P98175
Gene Alias DXS8237E; E430039K10Rik; G patch domain-containing protein 9; GPATC9; GPATCH9; Kiaa0122; RBM10; RNA binding motif protein 10; rna-binding; RNA-binding motif protein 10; RNA-binding protein 10; RNA-binding protein S1-1; S1-1; S1-1 protein from liver; TARPS; ZRANB5
Gene Symbols RBM10
Host Species Rabbit
Immunogen Recombinant Human RBM10. Recombinant protein control fragment (Product #RP-105070).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8241
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.