Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540331
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540331 Supplier Invitrogen™ Supplier No. PA540331
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QRRRRRRHRH SPTGPPGFPR DGDYRDQDYR TEQGEEEEEE EDEEEEEKAS Sequence homology: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

The protein encoded by this gene contains RNA recognition motif found in a variety of RNA binding proteins, including various hnRNP proteins, proteins implicated in regulation of alternative splicing, and protein components of snRNPs. In vitro studies showed that the rat homolog bound to RNA homopolymers, with a preference for G and U polyribonucleotides. This gene is part of a gene cluster on chromosome Xp11.23, and its 3' end lies within 20 kb upstream of UBE1. Two transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RBM10
Gene Accession No. P98175, Q99KG3
Gene Alias DXS8237E; E430039K10Rik; G patch domain-containing protein 9; GPATC9; GPATCH9; Kiaa0122; RBM10; RNA binding motif protein 10; rna-binding; RNA-binding motif protein 10; RNA-binding protein 10; RNA-binding protein S1-1; S1-1; S1-1 protein from liver; TARPS; ZRANB5
Gene Symbols RBM10
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RBM10 (aa 79-128).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 236732, 8241
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.