Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBM10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583253
Description
Immunogen sequence: MEYERRGGRG DRTGRYGATD RSQDDGGENR SRDHDYRDMD YRSYPREYGS QEGKHDYDDS SEEQSAEDSY EASPGSETQR R.
The protein encoded by this gene contains RNA recognition motif found in a variety of RNA binding proteins, including various hnRNP proteins, proteins implicated in regulation of alternative splicing, and protein components of snRNPs. In vitro studies showed that the rat homolog bound to RNA homopolymers, with a preference for G and U polyribonucleotides. This gene is part of a gene cluster on chromosome Xp11.23, and its 3' end lies within 20 kb upstream of UBE1. Two transcript variants encoding different isoforms have been identified for this gene.
Specifications
RBM10 | |
Polyclonal | |
Unconjugated | |
RBM10 | |
DXS8237E; E430039K10Rik; G patch domain-containing protein 9; GPATC9; GPATCH9; Kiaa0122; RBM10; RNA binding motif protein 10; rna-binding; RNA-binding motif protein 10; RNA-binding protein 10; RNA-binding protein S1-1; S1-1; S1-1 protein from liver; TARPS; ZRANB5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8241 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P98175 | |
RBM10 | |
Recombinant protein corresponding to Human RBM10. Recombinant protein control fragment (Product #RP-95471). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction