Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560440
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560440 Supplier Invitrogen™ Supplier No. PA560440
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NMIELSRRRF ETANLDIPPA NASRSGPPPS SGMSSRVNLP TTVSNFNNPS PSVVTATTSV HESNKNIQTF STASVGTAPP NMGASFGSPT FSSTVPST Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.

This gene encodes a protein that contains several RNA-binding motifs, potential transmembrane domains, and proline-rich regions. This gene and the gene for copine I overlap at map location 20q11.21. Alternative splicing in the 5' UTR results in two transcript variants. Both variants encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM12
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RBM12
Gene Accession No. Q8R4X3, Q9NTZ6
Gene Alias 5730420G12Rik; 9430070C08Rik; AI852903; Copine I; Copine-1; Cpne1; HRIHFB2091; KIAA0765; mKIAA0765; RBM12; RNA binding motif protein 12; RNA-binding motif protein 12; RNA-binding protein 12; SH3/WW domain anchor protein in the nucleus; SWAN
Gene Symbols RBM12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RBM12. Recombinant protein control fragment (Product #RP-99476).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10137, 652928, 75710
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.