Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110278 Supplier Invitrogen™ Supplier No. PA5110278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGPPGFGSGP PGLGSAPGHL GGPPAFGPGP GPGPGPGPIH IGGPPGFASS SGKPGPTVIK VQNMPFTVSI DEILDFFYGY QVIPGSVCLK YNEKGMPTGE AMVAFESRDE ATAAVIDLND RPIGSRKVKL VLG.

This gene encodes a protein that contains several RNA-binding motifs, potential transmembrane domains, and proline-rich regions. This gene and the gene for copine I overlap at map location 20q11.21. Alternative splicing in the 5' UTR results in two transcript variants. Both variants encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM12
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RBM12
Gene Accession No. Q9NTZ6
Gene Alias 5730420G12Rik; 9430070C08Rik; AI852903; Copine I; Copine-1; Cpne1; HRIHFB2091; KIAA0765; mKIAA0765; RBM12; RNA binding motif protein 12; RNA-binding motif protein 12; RNA-binding protein 12; SH3/WW domain anchor protein in the nucleus; SWAN
Gene Symbols RBM12
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 800 to the C-terminus of human RBM12 (NP_001185767.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10137
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.