Learn More
RBM12 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301198AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
This gene encodes a protein that contains several RNA-binding motifs, potential transmembrane domains, and proline-rich regions. This gene and the gene for copine I overlap at map location 20q11.21. Alternative splicing in the 5′ UTR results in two transcript variants. Both variants encode the same protein.Specifications
RBM12 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9NTZ6 | |
RBM12 | |
Between 625 and 675 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
RBM12 | |
RNA binding motif protein 12, HRIHFB2091, KIAA0765, SWAN | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10137 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.