Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM12B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565758
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565758 Supplier Invitrogen™ Supplier No. PA565758
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARRAISRSGGF IKDSSVELFL SSKAEMQKTI EMKRTDRVGR GRPGSGTSGV DSLSNFIESV KEEASNSGYG Highest antigen sequence identity to the following orthologs - mouse 80%, rat 75%.

RBM12B gene ontology annotations related to this gene include RNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM12B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RBM12B
Gene Accession No. Q8IXT5
Gene Alias 3000004N20Rik; MGC:33837; Rbm12b; Rbm12b1; Rbm12ba; RGD1307658; RNA binding motif protein 12 B1; RNA binding motif protein 12B; RNA-binding motif protein 12B; RNA-binding motif protein 12B-A; RNA-binding protein 12B; RNA-binding protein 12B-A
Gene Symbols RBM12B
Host Species Rabbit
Immunogen Recombinant Human RBM12B. Recombinant protein control fragment (Product #RP-106438).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389677
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.