Learn More
Invitrogen™ RBM12B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589109
Description
Immunogen sequence: KFASCVDASG GLKCHRSFMG SRFIEVMQGS EQQWIEFGGN AVKEGDVLRR SEEHSPPRGI NDRHFRKRSH SKSPRRTRSR SPLGFYVHLK NLSLSIDERD LRNFFRGTDL TDEQIRFLYK DENRTRYAFV MFKTLKDYNT ALSLHKTVLQ YRPVHIDPIS RKQMLKFIAR YEKKRSGSLE RDRPGHVSQK YSQEGNSGQK L; Positive Samples: HeLa.
RBM12B gene ontology annotations related to this gene include RNA binding.
Specifications
RBM12B | |
Polyclonal | |
Unconjugated | |
RBM12B | |
3000004N20Rik; MGC:33837; Rbm12b; Rbm12b1; Rbm12ba; RGD1307658; RNA binding motif protein 12 B1; RNA binding motif protein 12B; RNA-binding motif protein 12B; RNA-binding motif protein 12B-A; RNA-binding protein 12B; RNA-binding protein 12B-A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
389677 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.88 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IXT5 | |
RBM12B | |
Recombinant fusion protein containing a sequence corresponding to amino acids 200-400 of human RBM12B (NP_9763242). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.