Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM12B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | RBM12B |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1398
![]() ![]() |
Proteintech Group Inc
171371AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
RBM12B gene ontology annotations related to this gene include RNA binding.Specifications
RBM12B | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q80YR9, Q8IXT5 | |
313069, 389677, 72397 | |
RBM12B Fusion Protein Ag10894 | |
Primary | |
-20°C | |
RBM12B |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
RBM12B, RNA binding motif protein 12B, RNA binding protein 12B | |
Rbm12b, Rbm12b1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title