Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM14 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002884AF488
Description
RBM14 Polyclonal antibody specifically detects RBM14 in Human samples. It is validated for Western BlotSpecifications
RBM14 | |
Polyclonal | |
Western Blot | |
COAAMGC31756, Paraspeckle protein 2, PSP2DKFZp779J0927, RNA binding motif protein 14, RNA-binding motif protein 14, RNA-binding protein 14, RRM-containing coactivator activator/modulator, SIPMGC15912, Synaptotagmin-interacting protein, SYT-interacting protein, SYTIP1, TMEM137, transmembrane protein 137 | |
The immunogen recognized by this antibody maps to a region between residue 550 and 600 of human CoActivator Activator (RNA Binding Motif 14) using the numbering given in entry NP_006319.1 (GeneID 10432 ). | |
0.1 mL | |
Cell Cycle and Replication, DNA Repair | |
10432 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction