Learn More
Invitrogen™ RBM14 Polyclonal Antibody

Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (98%), Rat (98%), Pig (99%), Chimpanzee (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | RBM14 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | RBM14 |
| Gene Accession No. | Q96PK6 |
| Gene Alias | 1300007E16Rik; COAA; coactivator activator; p16; p16K; Paraspeckle protein 2; PSP2; Rbm14; RNA binding motif protein 14; rna-binding; RNA-binding motif protein 14; RNA-binding protein 14; RRM-containing coactivator activator/modulator; SIP; synaptotagmin interacting protein; synaptotagmin-interacting protein; SYT-interacting protein; Sytip; SYTIP1; TMEM137; transmembrane protein 137; Unknown (protein for MGC:137891) |
| Gene Symbols | RBM14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.