Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552314
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552314 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552314 Supplier Invitrogen™ Supplier No. PA552314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TQSSASLAAS YAAQQHPQAA ASYRGQPGNA YDGAGQPSAA YLSMSQGAVA NANSTPPPYE RTRLSPPRAS YDDPYKKAVA MSKRYGSDRR LAELSDYRRL SESQLSFRRS PTKSSLDYRR LPDAHSDYAR Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Isoform 1 may function as a nuclear receptor coactivator, enhancing transcription through other coactivators such as NCOA6 and CITED1. Isoform 2, functions as a transcriptional repressor, modulating transcriptional activities of coactivators including isoform 1, NCOA6 and CITED1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM14
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RBM14
Gene Accession No. Q8C2Q3, Q96PK6
Gene Alias 1300007E16Rik; COAA; coactivator activator; p16; p16K; Paraspeckle protein 2; PSP2; Rbm14; RNA binding motif protein 14; rna-binding; RNA-binding motif protein 14; RNA-binding protein 14; RRM-containing coactivator activator/modulator; SIP; synaptotagmin interacting protein; synaptotagmin-interacting protein; SYT-interacting protein; Sytip; SYTIP1; TMEM137; transmembrane protein 137; Unknown (protein for MGC:137891)
Gene Symbols RBM14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RBM14. Recombinant protein control fragment (Product #RP-89418).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10432, 56275
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.