Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589036
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589036 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589036 Supplier Thermo Scientific Supplier No. PA589036
Add to Cart
Add to Cart

RBM14 Polyclonal Antibody

RBM14 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM14
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RBM14
Gene Accession No. Q8C2Q3, Q96PK6
Gene Alias 1300007E16Rik; COAA; coactivator activator; p16; p16K; Paraspeckle protein 2; PSP2; Rbm14; RNA binding motif protein 14; rna-binding; RNA-binding motif protein 14; RNA-binding protein 14; RRM-containing coactivator activator/modulator; SIP; synaptotagmin interacting protein; synaptotagmin-interacting protein; SYT-interacting protein; Sytip; SYTIP1; TMEM137; transmembrane protein 137; Unknown (protein for MGC:137891)
Gene Symbols RBM14
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 110-220 of human RBM14 (NP_0063191).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10432, 170900, 56275
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.