Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM15 Mouse anti-Human, Clone: 4A1A4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | RBM15 |
---|---|
Clone | 4A1A4 |
Concentration | 0.99 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6498
![]() ![]() |
Proteintech Group Inc
660591IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
May function as an mRNA export factor, stimulating export and expression of RTE-containing mRNAs which are present in many retrotransposons that require to be exported prior to splicing. High affinity binding of pre-mRNA to RBM15 may allow targeting of the mRNP to the export helicase DBP5 in a manner that is independent of splicing-mediated NXF1 deposition, resulting in export prior to splicing. May be implicated in HOX gene regulation.Specifications
RBM15 | |
0.99 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
One twenty two protein 1, OTT, OTT1, RBM15, RNA binding motif protein 15, SPEN | |
RBM15 | |
IgG2a | |
Protein A | |
Antibody |
4A1A4 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q96T37 | |
64783 | |
RBM15 Fusion Protein Ag17970 | |
Primary | |
-20°C | |
RBM15 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title