Learn More
Invitrogen™ RBM15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592321
Description
Immunogen sequence: EHRYQQQYLQ PLPLTHYELV TDAFGHRAPD PLRGARDRTP PLLYRDRDRD LYPDSDWVPP PPPVRERSTR TAATSVPAYE PLDSLDRRRD GWSLDRDRGD RDLPSSRDQP RKRRLPEESG GRHLDRSPES DRPRKRHCAP SPDRSPELSS SRDRYNSDND RSSRLLLERP SPIRDRRGSL EKSQGDKRDR KNSASAERDR KHRTTAPTEG KSPLKKEDRS DGSAPSTSTA SSKLKSPSQK QDGGTAPVAS A; Positive Samples: HL-60, HepG2, 293T, Jurkat, Mouse thymus, Mouse spleen; Cellular Location: Nucleus, Nucleus membrane, Nucleus speckle, Peripheral membrane protein, nucleoplasm.
May function as an mRNA export factor, stimulating export and expression of RTE-containing mRNAs which are present in many retrotransposons that require to be exported prior to splicing. High affinity binding of pre-mRNA to RBM15 may allow targeting of the mRNP to the export helicase DBP5 in a manner that is independent of splicing-mediated NXF1 deposition, resulting in export prior to splicing. May be implicated in HOX gene regulation.
Specifications
RBM15 | |
Polyclonal | |
Unconjugated | |
RBM15 | |
C230088J01Rik; Kiaa4257; mKIAA1438; one twenty two protein; one twenty-two; one-twenty two protein 1; OTT; OTT1; putative RNA-binding protein 15; RBM15; RNA binding motif protein 15; RNA-binding motif protein 15; RNA-binding protein 15; SPEN | |
Rabbit | |
Affinity chromatography | |
RUO | |
229700, 64783 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96T37 | |
RBM15 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 530-780 of human RBM15 (NP_0011884741). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.