Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBM15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599993
Description
Antibody detects endogenous levels of total RBM15.
May function as an mRNA export factor, stimulating export and expression of RTE-containing mRNAs which are present in many retrotransposons that require to be exported prior to splicing. High affinity binding of pre-mRNA to RBM15 may allow targeting of the mRNP to the export helicase DBP5 in a manner that is independent of splicing-mediated NXF1 deposition, resulting in export prior to splicing. May be implicated in HOX gene regulation.
Specifications
RBM15 | |
Polyclonal | |
Unconjugated | |
RBM15 | |
C230088J01Rik; Kiaa4257; mKIAA1438; one twenty two protein; one twenty-two; one-twenty two protein 1; OTT; OTT1; putative RNA-binding protein 15; RBM15; RNA binding motif protein 15; RNA-binding motif protein 15; RNA-binding protein 15; SPEN | |
Rabbit | |
Affinity chromatography | |
RUO | |
229700, 64783, 684233 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96T37 | |
RBM15 | |
A synthesized peptide derived from human RBM15(Accession Q96T37), corresponding to amino acid residues M45-R95. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction