Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM15B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501733796 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3796 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3796 Supplier Proteintech Group Inc Supplier No. 222491AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This RBM15B antibody was produced by immunizing animals with the C-terminus of RBM15B, which is homologous to a short form of RBM15B (563 aa, ∽70 kDa), thus this antibody can recognize full length of RBM15B (∽100 kDa) and splice-one (70 kDa).

May function in the regulation of alternative or illicit splicing.

Specifications

Antigen RBM15B
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RBM15B
Gene Accession No. Q6PHZ5, Q8NDT2
Gene Alias HUMAGCGB, HuOTT3, One twenty two protein 3, OTT3, RBM15B, RNA binding motif protein 15B
Gene Symbols Rbm15b
Host Species Rabbit
Immunogen RBM15B Fusion Protein Ag17744
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109095, 29890, 315988
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.