Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBM17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557964
Description
Immunogen sequence: GLGKHEQGLS TALSVEKTSK RGGKIIVGDA TEKDASKKSD SNPLTEILKC PTKVVLLRNM VGAGEVDEDL EVETKEECEK YGKVGKCVIF EIPGAPDDEA VRIF Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
This gene encodes an RNA binding protein. The encoded protein is part of the spliceosome complex and functions in the second catalytic step of mRNA splicing. Alternatively spliced transcript variants have been described. Related pseudogenes exist on chromosomes 9 and 15.
Specifications
| RBM17 | |
| Polyclonal | |
| Unconjugated | |
| Rbm17 | |
| 2700027J02Rik; 45 kDa-splicing factor; Rbm17; RNA binding motif protein 17; RNA-binding motif protein 17; Spf45; Splicing factor 45; splicing factor 45kDa | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 291295, 76938, 84991 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8JZX4, Q96I25 | |
| Rbm17 | |
| Recombinant protein corresponding to Human RBM17. Recombinant protein control fragment (Product #RP-97079). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction