Learn More
Invitrogen™ RBM17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA591967
Description
Immunogen sequence: MSLYDDLGVE TSDSKTEGWS KNFKLLQSQL QVKKAALTQA KSQRTKQSTV LAPVIDLKRG GSSDDRQIVD TPPHVAAGLK DPVPSGFSAG EVLIPLADEY DPMFPNDYEK VVKRQREERQ RQRELERQKE IEEREKRRKD RHEASGFARR PDPDSDEDED YERERRKRSM GGAAIAPPTS LVEKDKELPR DFPYEEDSRP RSQSSKAAIP PPVYEEQDRP RSPTGPSNSF LANMGGTVAH; Positive Samples: LO2, A-549, HeLa, DU145, Rat thymus; Cellular Location: Nucleus.
This gene encodes an RNA binding protein. The encoded protein is part of the spliceosome complex and functions in the second catalytic step of mRNA splicing. Alternatively spliced transcript variants have been described. Related pseudogenes exist on chromosomes 9 and 15.
Specifications
RBM17 | |
Polyclonal | |
Unconjugated | |
Rbm17 | |
2700027J02Rik; 45 kDa-splicing factor; Rbm17; RNA binding motif protein 17; RNA-binding motif protein 17; Spf45; Splicing factor 45; splicing factor 45kDa | |
Rabbit | |
Affinity Chromatography | |
RUO | |
291295, 76938, 84991 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8JZX4, Q96I25 | |
Rbm17 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human RBM17 (NP_1162941). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.