Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598774
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598774 Supplier Invitrogen™ Supplier No. PA598774
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RBM17 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes an RNA binding protein. The encoded protein is part of the spliceosome complex and functions in the second catalytic step of mRNA splicing. Alternatively spliced transcript variants have been described. Related pseudogenes exist on chromosomes 9 and 15.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM17
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Rbm17
Gene Accession No. Q96I25
Gene Alias 2700027J02Rik; 45 kDa-splicing factor; Rbm17; RNA binding motif protein 17; RNA-binding motif protein 17; Spf45; Splicing factor 45; splicing factor 45kDa
Gene Symbols Rbm17
Host Species Rabbit
Immunogen Recombinant Human Splicing factor 45 protein (1-240AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84991
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.