Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558567
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558567 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558567 Supplier Invitrogen™ Supplier No. PA558567
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKNTTKSWQG RILGENEEEE DLAESGRLFV RNLPYTSTEE DLEKLFSKYG PLSELHYPID SLTKKPKGFA FITFMFPEHA VKAYSEVDGQ VFQGRMLHVL PSTIKKEASE Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 86%.

Plays a role in embryo pre-implantation development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM19
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RBM19
Gene Accession No. Q8R3C6, Q9Y4C8
Gene Alias 1200009A02Rik; AV302714; KIAA0682; NPO; probable RNA-binding protein 19; Rbm19; RNA binding motif protein 19; RNA-binding motif protein 19
Gene Symbols RBM19
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RBM19. Recombinant protein control fragment (Product #RP-97717).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304512, 74111, 9904
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.