Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rbm20 Polyclonal Antibody

Catalog No. PIPA5121650
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121650 20 μL
PIPA5119402 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121650 Supplier Invitrogen™ Supplier No. PA5121650
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Rbm20 Polyclonal Antibody for Western Blot

This gene encodes a protein that likely binds RNA. Mutations in this gene have been associated with familial dilated cardiomyopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rbm20
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Rbm20
Gene Accession No. Q5T481
Gene Alias 1110018J23Rik; 2010003H22Rik; AI646761; probable RNA-binding protein 20; Rbm20; RNA binding motif protein 20; RNA-binding motif protein 20; RNA-binding protein 20
Gene Symbols Rbm20
Host Species Rabbit
Immunogen Recombinant protein RNA Binding Motif Protein 20. The antigen corresponds to amino acid range 1089-1227 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100157563, 282996
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.