Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541877
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541877 Supplier Invitrogen™ Supplier No. PA541877
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KWGRSQAARG KEKEKDGTTD SGIKLEPVPG LPGALPPPPA AEEEASANYF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

This gene encodes an RNA binding protein. The encoded protein may play a role in cell division and may be involved in pre-mRNA splicing. Related pseudogenes exist on chromosomes 6, 7, 9, 13, 16, 18, and X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM22
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RBM22
Gene Accession No. Q9NW64
Gene Alias 199G4; 8430430L24Rik; Cwc2; fSAP47; functional spliceosome-associated protein 47; pre-mRNA-splicing factor RBM22; RBM22; RNA binding motif protein 22; RNA-binding motif protein 22; Unknown (protein for MGC:128706); ZC3H16; Zinc finger CCCH domain-containing protein 16
Gene Symbols RBM22
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human RBM22 (aa 301-350).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55696
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.