Learn More
RBM22 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303924AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Chicken, Bovine and Orangutan
This gene encodes an RNA binding protein. The encoded protein may play a role in cell division and may be involved in pre-mRNA splicing. Related pseudogenes exist on chromosomes 6, 7, 9, 13, 16, 18, and X.Specifications
RBM22 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8BHS3, Q9NW64 | |
Rbm22 | |
Between 370 and 420 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RBM22 | |
Cwc2, ZC3H16, fSAP47, RNA-binding motif protein 22, functional spliceosome-associated protein 47, pre-mRNA-splicing factor RBM22, zinc finger CCCH domain-containing protein 16 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55696, 66810 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.