Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117673
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117673 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117673 Supplier Invitrogen™ Supplier No. PA5117673
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

RBM24 is preferentially expressed in muscle during differentiation in vitro and may regulate myogenic differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM24
Applications Western Blot
Classification Polyclonal
Concentration 4.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene rbm24a
Gene Accession No. Q76LC6
Gene Alias 6330546B05Rik; AI606861; dJ259A10.1; Rbm24; rbm24a; RNA binding motif protein 24; RNA binding motif protein 24a; RNA binding protein; rna-binding; RNA-binding motif protein 24; RNA-binding protein 24; RNA-binding protein SEB-4; RNA-binding region (RNP1, RRM) containing 6; RNA-binding region-containing protein 6; RNPC6; seb-4
Gene Symbols rbm24a
Host Species Rabbit
Immunogen Recombinant Zebrafish RNA-binding protein 24 protein (2-78aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 405801
Target Species Zebrafish
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.