Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM25 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | RBM25 |
---|---|
Dilution | Western Blot 0.04 - 0.4 ug/ml, Simple Western 1:25, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
RBM25 Polyclonal specifically detects RBM25 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RBM25 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
58517 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:PVDSVFNKFEDEDSDDVPRKRKLVPLDYGEDDKNATKGTVNTEEKRKHIKSLIEKIPTAKPELFAYPLDWSIVDSILMERRIRPWINKKIIEYIGEEEATLVDFVCSKVMAHSSPQSILDDVAMVLDEEAEV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04 - 0.4 ug/ml, Simple Western 1:25, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Human | |
Arg/Glu/Asp-rich protein of 120 kDa, Arg/Glu/Asp-rich protein, 120 kDa, fSAP94, functional spliceosome-associated protein 94, MGC105088, MGC117168, NET52, Protein S164, RED120RNA-binding region-containing protein 7, RNA binding motif protein 25, RNA-binding motif protein 25, RNA-binding protein 25, RNA-binding region (RNP1, RRM) containing 7, RNPC7, S164, Snu71, U1 small nuclear ribonucleoprotein 1SNRP homolog | |
RBM25 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title