Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590511
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590511 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590511 Supplier Invitrogen™ Supplier No. PA590511
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KPTLRPISSA PSVSSASGNA TPNTPGDESP CGIIIPHENS PDQQQPEEHR PKIGLSLKLG ASNSPGQPNS VKRKKLPVDS VFNKFEDEDS DDVPRKRKLV PLDYGEDDKN ATKGTVNTEE KRKHIKSLIE KIPTAKPELF AYPLDWSIVD SILMERRIRP WINKKIIEYI GEEEATLVDF VCSKVMAHSS PQSILDDVAM VLDEEAEVFI VKMWRLLIYE TEAKKIGLVK; Positive Samples: DU145, OVCAR3, HepG2, Jurkat, NIH/3T3, mouse brain, mouse liver; Cellular Location: Cytoplasm, Nucleus speckle.

RNA-binding protein that acts as a regulator of alternative pre-mRNA splicing. Involved in apoptotic cell death through the regulation of the apoptotic factor BCL2L1 isoform expression. Modulates the ratio of proapoptotic BCL2L1 isoform S to antiapoptotic BCL2L1 isoform L mRNA expression. When overexpressed, stimulates proapoptotic BCL2L1 isoform S 5'-splice site (5'-ss) selection, whereas its depletion caused the accumulation of antiapoptotic BCL2L1 isoform L. Promotes BCL2L1 isoform S 5'-ss usage through the 5'-CGGGCA-3' RNA sequence. Its association with LUC7L3 promotes U1 snRNP binding to a weak 5' ss in a 5'-CGGGCA-3'-dependent manner. Binds to the exonic splicing enhancer 5'-CGGGCA-3' RNA sequence located within exon 2 of the BCL2L1 pre-mRNA. Also involved in the generation of an abnormal and truncated splice form of SCN5A in heart failure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM25
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RBM25
Gene Accession No. B2RY56, P49756
Gene Alias 2600011C06Rik; 2610015J01Rik; A130095G20Rik; AI159652; AL023075; arg/Glu/Asp-rich protein of 120 kDa; Arg/Glu/Asp-rich protein, 120 kDa; AU043498; fSAP94; functional spliceosome-associated protein 94; LOC540258 protein; NET52; Protein S164; Rbm25; RED120; RNA binding motif protein 25; rna-binding; RNA-binding motif protein 25; RNA-binding protein 25; RNA-binding protein 25-like protein; RNA-binding region (RNP1, RRM) containing 7; RNA-binding region-containing protein 7; RNPC7; S164; Snu71; U1 small nuclear ribonucleoprotein 1SNRP homolog
Gene Symbols RBM25
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 614-843 of human RBM25 (NP_0670621).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58517, 67039
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.