Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531667
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531667 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531667 Supplier Invitrogen™ Supplier No. PA531667
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, HepG2, HCT116. Predicted reactivity: Mouse (92%), Rat (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RBM26 (RNA Binding Motif Protein 26) is a Protein Coding gene. Diseases associated with RBM26 include Cutaneous T Cell Lymphoma and Helsmoortel-Van Der Aa Syndrome. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and nucleotide binding. An important paralog of this gene is RBM27. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM26
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene RBM26
Gene Accession No. Q5T8P6
Gene Alias 1700009P03Rik; acidic rich RS domain containing 2; AI447318; ARRS2; C13orf10; C230097K14Rik; C86225; CTCL tumor antigen se70-2; cutaneous T-cell lymphoma tumor antigen se70-2; fc66d05; MLZ-393; PPP1R132; Pro1777; protein expressed in male leptotene and zygotene spermatocytes 393; protein phosphatase 1, regulatory 132; Rbm26; RGD1308297; RNA binding motif protein 26; RNA-binding motif protein 26; RNA-binding protein 26; RP11-255E21.1; SE70-2; wu:fc66d05; ZC3H17; zgc:109736
Gene Symbols RBM26
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RBM26. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64062
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.