Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555371
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555371 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555371 Supplier Invitrogen™ Supplier No. PA555371
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EHSKGCAFAQ FMTQEAAQKC LLAASPENEA GGLKLDGRQL KVDLAVTRDE AAKLQTTKVK KPTGTRNLYL AREGLIRAGT KAAEGVSAAD MAKRERFELL KHQKLKDQNI FVSRTR Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

Phosphoinositide-binding protein which associates with both cell and endoplasmic reticulum (ER) membranes. Can bind to the ER membrane protein VAPA and recruit VAPA to plasma membrane sites, thus linking these intracellular compartments. The ORP3-VAPA complex stimulates RRAS signaling which in turn attenuates integrin beta-1 (ITGB1) activation at the cell surface. With VAPA, may regulate ER morphology. Has a role in regulation of the actin cytoskeleton, cell polarity and cell adhesion. Binds to phosphoinositides with preference for PI(3,4)P2 and PI(3,4,5)P3. Also binds 25-hydroxycholesterol and cholesterol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM28
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rbm28
Gene Accession No. Q8CGC6, Q9NW13
Gene Alias 2810480G15Rik; AI503051; ANES; hypothetical protein MGC56258; RBM28; RNA binding motif protein 28; RNA-binding motif protein 28; RNA-binding protein 28; zgc:56258
Gene Symbols Rbm28
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RBM28. Recombinant protein control fragment (Product #RP-92673).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 312182, 55131, 68272
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.