Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP243829
Description
Specifically detects RBM3 in Human samples, and it is validated for Immunocytochemistry, Immunofluorescence
RBM3 Polyclonal specifically detects RBM3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
RBM3 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
IS1-RNPL, putative RNA-binding protein 3, RNA binding motif (RNP1, RRM) protein 3, RNA binding motif protein 3, RNA-binding motif protein 3, RNPL | |
Rabbit | |
17 kDa | |
0.1 mL | |
metabolism | |
5935 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
RBM3 | |
Recombinant protein encompassing a sequence within the center region of human RBM3. The exact sequence is proprietary. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction