Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM3 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297785PCP
Description
RBM3 Polyclonal antibody specifically detects RBM3 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
RBM3 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
IS1-RNPL, putative RNA-binding protein 3, RNA binding motif (RNP1, RRM) protein 3, RNA binding motif protein 3, RNA-binding motif protein 3, RNPL | |
Produced in rabbits immunized with E. coli-derived Human RBM3 fragment. | |
0.1 mL | |
metabolism | |
5935 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction