Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551976 Supplier Invitrogen™ Supplier No. PA551976
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DEQALEDHFS SFGPISEVVV VKDRETQRSR GFGFITFTNP EHASVAMRAM NGESLDGRQI RVDHAGKSAR GTRGGGFGAH GRGRSYSRGG GDQGYGSGRY YDSRPGGYGY GYGRSRDYNG RNQGGYDRYS GGNY Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 96%.

RBM3 is a member of the glycine-rich RNA-binding protein family and a protein with one RNA recognition motif (RRM) domain. Expression of this protein is induced by cold shock and low oxygen tension.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RBM3
Gene Accession No. O89086, P98179, Q925G0
Gene Alias 2600016C11Rik; hypothetical protein LOC509771; IS1-RNPL; OTTHUMP00000025800; OTTHUMP00000025802; putative RNA-binding protein 3; Rbm3; RNA binding motif (RNP1, RRM) protein 3; RNA binding motif protein 3; RNA-binding motif protein 3; RNA-binding protein 3; RNPL
Gene Symbols RBM3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RBM3. Recombinant protein control fragment (Product #RP-92189).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114488, 19652, 5935
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.