Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 143631AP150UL

Description
This gene is a member of the glycine-rich RNA-binding protein family and encodes a protein with one RNA recognition motif domain. Expression of this gene is induced by cold shock and low oxygen tension. A pseudogene exists on chromosome 1. Multiple alternatively spliced transcript variants that are predicted to encode different isoforms have been characterized although some of these variants fit nonsense-mediated decay criteria.Specifications
RBM3 | |
Polyclonal | |
Unconjugated | |
RBM3 | |
IS1 RNPL, Putative RNA binding protein 3, RBM3, RNA binding motif protein 3, RNPL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114488, 19652, 5935 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O89086, P98179, Q925G0 | |
RBM3 | |
RBM3 Fusion Protein Ag5934 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction