Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM3 Recombinant Rabbit Monoclonal Antibody (JE63-84)
GREENER_CHOICE

Catalog No. PIMA544679
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544679 Supplier Invitrogen™ Supplier No. MA544679
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

RBM3 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Flow

RBM3 is a member of the glycine-rich RNA-binding protein family and a protein with one RNA recognition motif (RRM) domain. Expression of this protein is induced by cold shock and low oxygen tension.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE63-84
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene RBM3
Gene Accession No. O89086, P98179, Q925G0
Gene Alias 2600016C11Rik; hypothetical protein LOC509771; IS1-RNPL; OTTHUMP00000025800; OTTHUMP00000025802; putative RNA-binding protein 3; Rbm3; RNA binding motif (RNP1, RRM) protein 3; RNA binding motif protein 3; RNA-binding motif protein 3; RNA-binding protein 3; RNPL
Gene Symbols RBM3
Host Species Rabbit
Immunogen Recombinant protein within human RBM3 aa 58-157/157.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114488, 19652, 5935
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.