Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM33 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222278IR
Description
RBM33 Polyclonal antibody specifically detects RBM33 in Human, Mouse samples. It is validated for Western BlotSpecifications
RBM33 | |
Polyclonal | |
Western Blot | |
DKFZp434D1319, MGC20460, proline-rich protein 8, RNA binding motif protein 33, RNA-binding motif protein 33, RNA-binding protein 33 | |
The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human RNA Binding Motif Protein 33 using the numbering given in entry NP_444271.2 (GeneID 155435). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
155435 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction